N-Succinimidyl 6-Biotinamidohexanoate
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 6-Biotinamidohexanoic Acid N-Succinimidyl Ester, N-Biotinyl-6-aminohexanoic Acid Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl 6-(5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)hexanoate, N-[5-(N-Succinimidyloxycarbonyl)pentyl]biotinamide, Biotin-LC-NHS, Biotin-C5-NHS Ester
CAS Number:72040-63-2
:
Formula:C₂₀H₃₀N₄O₆S
MDL Number:MFCD00065502
:454.54 g/mol
Melting Pt:169 °C
:GHS07
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together