
1-Aminocyclopropanecarboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:22059-21-8
:
Formula:C₄H₇NO₂
MDL Number:MFCD00009944
:101.11 g/mol
EINECS:606-917-8
:GHS07
Storage Temperature:Ambient
Frequently Bought Together