Order Entry
Czech Republic
ContactUsLinkComponent
5-Oxa-2-azaspiro[3.4]octane oxalate (2:1)
5-Oxa-2-azaspiro[3.4]octane oxalate (2:1)
Catalog # APOSOR317208-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Oxa-2-azaspiro[3.4]octane oxalate (2:1)
Catalog # APOSOR317208-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :316.36 g/mol
Storage Temperature:Ambient
CAS Number:1523618-29-2
Formula:C₁₄H₂₄N₂O₆
MDL Number:MFCD27988000