Human recombinant Tau-441 (from E. coli)
Catalog # ACRBTAU-H51H3100UG
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Pk:100 µG
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:01
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Gene ID:P10636-8
- Protein synonyms:FTDP-17|MAPT|TAU|MTBT1|PHF-tau|MSTD|Tau|DDPAC
- Protein/peptide name:Tau-441
- Purity:>90% as determined by reduced SDS-PAGE.
- Molecular weight:47,9 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, 1 mM TCEP, 1 mM EDTA, pH7,5. Normally trehalose is added as protectant before lyophilisation
- Shipping temperature:RT
Specifications
About this item
Human Tau-441 / 2N4R Protein Tag.