Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant TGF-beta 1 (from HEK293 cells), Biotin
Human recombinant TGF-beta 1 (from HEK293 cells), Biotin
Catalog # ACRBTG1-H8217-20UG
Supplier:  ACROBIOSYSTEMS
CAS Number:  
Human recombinant TGF-beta 1 (from HEK293 cells), Biotin
Catalog # ACRBTG1-H8217-20UG
Supplier:  ACROBIOSYSTEMS
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Conjugation:
    Biotin
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Seq:
    01
  • Tag sequence:
    Avitag™
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Biological activity:
    Biotinylated Human TGF-Beta 1, Avitag™;;;;;;;
  • Gene ID:
    P01137-1
  • Protein synonyms:
    TGF-beta-1|CEDLAP|TGF-beta 1|TGFB|DPD1|TGFB1
  • Protein/peptide name:
    TGF-beta 1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    14,5 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 60% ACN, 0,085% TFA in Water. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

About this item

Biotinylated Human TGF-Beta 1 / TGFB1 Protein, Avitag™.