
4-Hydroxy-2-(pyrazin-2-yl)-6-(trifluoromethyl)pyrimidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:438249-86-6
:
Formula:C₉H₅F₃N₄O
MDL Number:MFCD00665787
:242.157 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together