0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
6-Hydroxy-4-(trifluoromethyl)nicotinic acid
6-Hydroxy-4-(trifluoromethyl)nicotinic acid 95+%
6-Hydroxy-4-(trifluoromethyl)nicotinic acid 95+%
APOSPC9700-1G
:
Apollo Scientific
:
6-Hydroxy-4-(trifluoromethyl)nicotinic acid 95+%
APOSPC9700-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
207.107 g/mol
:
302…305 °C (dec.)
:
GHS07
:
Ambient
:
849020-87-7
:
C₇H₄F₃NO₃
:
MFCD06245493