N-Methyl-N-{2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}methylamine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:857284-26-5
:
Formula:C₁₂H₁₁F₃N₂S
MDL Number:MFCD07772873
:272.289 g/mol
Storage Temperature:Ambient
Frequently Bought Together