
4-Hydroxy-2-phenyl-6-(trifluoromethyl)pyrimidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:158715-14-1
:
Formula:C₁₁H₇F₃N₂O
MDL Number:MFCD00269571
:240.181 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together