
2-(3-Fluorophenyl)amino-4-(2-hydroxyphenyl)-1,3-thiazole hydrobromide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1209843-32-2
:
Formula:C₁₅H₁₁FN₂OS·BrH
MDL Number:MFCD08458146
:367.236 g/mol
Melting Pt:224…227 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together