Order Entry
Czech Republic
ContactUsLinkComponent
Methyl 3-{[2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-2-methylhydrazino]carbonyl}isonicotinate
Methyl 3-{[2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-2-methylhydrazino]carbonyl}isonicotinate
Catalog # APOSPC9401-500G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 3-{[2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-2-methylhydrazino]carbonyl}isonicotinate
Catalog # APOSPC9401-500G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :388.729 g/mol
Boiling Pt:168…170 °C
Melting Pt:168…170 °C
Storage Temperature:Ambient
CAS Number:338415-40-0
Formula:C₁₅H₁₂ClF₃N₄O₃
MDL Number:MFCD00975038