
2-(2,6-Difluorophenyl)amino-4-(2-hydroxyphenyl)-1,3-thiazole hydrobromide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1209726-22-6
:
Formula:C₁₅H₁₀F₂N₂OS·BrH
MDL Number:MFCD08458135
:385.226 g/mol
Melting Pt:>240 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together