
2-(4-Bromo-2-fluorophenyl)cyclopropane-1-carboxylic acid
Catalog # APOSPC9302-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:259.072 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1056010-27-5
Formula:C₁₀H₈BrFO₂
MDL Number:MFCD12068280