Ethyl (2E)-3-(2-fluoropyridin-3-yl)prop-2-enoate 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1563741-43-4
:
Formula:C₁₀H₁₀FNO₂
MDL Number:MFCD26711919
:195.19 g/mol
Storage Temperature:Ambient
Frequently Bought Together