Order Entry
Czech Republic
ContactUsLinkComponent
4-Fluoro-3-hydroxy-n,n-dimethylbenzamide 95%
4-Fluoro-3-hydroxy-n,n-dimethylbenzamide 95%
Catalog # APOSPC910305-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Fluoro-3-hydroxy-n,n-dimethylbenzamide 95%
Catalog # APOSPC910305-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :183.18 g/mol
Storage Temperature:Ambient
Formula:C₉H₁₀FNO₂
MDL Number:MFCD16997893