Order Entry
Czech Republic
ContactUsLinkComponent
 
3-Hydroxy-2-methoxy-5-(trifluoromethyl)pyridine 95%
Catalog # APOSPC910276-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Hydroxy-2-methoxy-5-(trifluoromethyl)pyridine 95%
Catalog # APOSPC910276-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.123 g/mol
Storage Temperature:Ambient
CAS Number:1227580-74-6
Formula:C₇H₆F₃NO₂
MDL Number:MFCD16611384