Order Entry
Czech Republic
ContactUsLinkComponent
2-Fluoro-3-(hydroxymethyl)phenol 95%
2-Fluoro-3-(hydroxymethyl)phenol 95%
Catalog # APOSPC910231-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Fluoro-3-(hydroxymethyl)phenol 95%
Catalog # APOSPC910231-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :142.128 g/mol
Storage Temperature:Ambient
CAS Number:960001-66-5
Formula:C₇H₇FO₂
MDL Number:MFCD12923294