Methyl 3-methyl-5-(trifluoromethyl)benzoate
Catalog # APOSPC909774-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:218.172 g/mol
Storage Temperature:Ambient
CAS Number:161622-18-0
Formula:C₁₀H₉F₃O₂
MDL Number:MFCD14698072