Order Entry
Czech Republic
ContactUsLinkComponent
1-(3',4'-Difluorophenyl)ethylamine 97%
1-(3',4'-Difluorophenyl)ethylamine 97%
Catalog # APOSPC908669-5G
Supplier:  Apollo Scientific
CAS Number:  
1-(3',4'-Difluorophenyl)ethylamine 97%
Catalog # APOSPC908669-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :157.161 g/mol
Storage Temperature:Ambient
CAS Number:276875-21-9
Formula:C₈H₉F₂N
MDL Number:MFCD04116333