6,6-Difluorospiro[3.3]heptan-2-amine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1423032-71-6
:
Formula:C₇H₁₁F₂N·ClH
MDL Number:MFCD22566009
:183.627 g/mol
Storage Temperature:Ambient
Frequently Bought Together