1-(2-Fluoro-4,5-dimethoxyphenyl)ethan-1-one 95%
Catalog # APOSPC908039-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:198.191 g/mol
Storage Temperature:Ambient
CAS Number:142265-69-8
Formula:C₁₀H₁₁FO₃
MDL Number:MFCD17226285