Order Entry
Czech Republic
ContactUsLinkComponent
4-Fluoropiperidine-4-carbonitrile hydrochloride ≥98%
4-Fluoropiperidine-4-carbonitrile hydrochloride ≥98%
Catalog # APOSPC906999-100MG
Supplier:  Apollo Scientific
CAS Number:  
4-Fluoropiperidine-4-carbonitrile hydrochloride ≥98%
Catalog # APOSPC906999-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :164.608 g/mol
Storage Temperature:Ambient
CAS Number:1374653-45-8
Formula:C₆H₉FN₂·ClH
MDL Number:MFCD19689455