
Boc-(4R)-4-(4-trifluoromethylbenzyl)-L-proline 98+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:957311-17-0
:
Formula:C₁₈H₂₂F₃NO₄
MDL Number:MFCD06659430
Melting Pt:150…152 °C
:373.367 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together