Order Entry
Czech Republic
ContactUsLinkComponent
2-Fluoro-5-hydroxymethyl-4-methylpyridine 95%
2-Fluoro-5-hydroxymethyl-4-methylpyridine 95%
Catalog # APOSPC904307-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Fluoro-5-hydroxymethyl-4-methylpyridine 95%
Catalog # APOSPC904307-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :141.143 g/mol
Storage Temperature:Ambient
CAS Number:1394899-05-8
Formula:C₇H₈FNO
MDL Number:MFCD22392285