
4-Bromo-2-fluoro-6-methoxybenzaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:856767-09-4
:
Formula:C₈H₆BrFO₂
MDL Number:MFCD22543644
:233.034 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together