6-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1026962-68-4
:
Formula:C₈H₃BrF₄O₂
MDL Number:MFCD21365092
:287.006 g/mol
Storage Temperature:Refrigerator
Frequently Bought Together