(1-Methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl)methanol 95%
Catalog # APOSPC903884-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:180.128 g/mol
Storage Temperature:Ambient
CAS Number:540468-96-0
Formula:C₆H₇F₃N₂O
MDL Number:MFCD09040887