Order Entry
Czech Republic
ContactUsLinkComponent
5-Cyano-2-fluorobenzene-1-sulphonyl chloride 95%
Corrosive
5-Cyano-2-fluorobenzene-1-sulphonyl chloride 95%
Catalog # APOSPC903871-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
5-Cyano-2-fluorobenzene-1-sulphonyl chloride 95%
Catalog # APOSPC903871-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :219.621 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
 :GHS05
CAS Number:1101120-80-2
Formula:C₇H₃ClFNO₂S
MDL Number:MFCD18394026