
3,3-Difluoro-3-(heptafluoropropyl)-1,2-propenoxide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:89807-87-4
:
Formula:C₆H₃F₉O
MDL Number:MFCD00042341
:262.073 g/mol
Boiling Pt:52 °C (41 mm Hg)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together