Order Entry
Czech Republic
ContactUsLinkComponent
1-Fluoro-3-iodo-2-methoxybenzene 95%
1-Fluoro-3-iodo-2-methoxybenzene 95%
Catalog # APOSPC903701-5G
Supplier:  Apollo Scientific
CAS Number:  
1-Fluoro-3-iodo-2-methoxybenzene 95%
Catalog # APOSPC903701-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :252.025 g/mol
Storage Temperature:Ambient
CAS Number:32750-21-3
Formula:C₇H₆FIO
MDL Number:MFCD07782083