
4-Hydroxy-2-(trifluoromethyl)benzaldehyde
Catalog # APOSPC903683-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:190.119 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1243395-68-7
Formula:C₈H₅F₃O₂
MDL Number:MFCD15528830