5-Bromo-3-fluoro-2-nitrophenylacetonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1807211-69-3
:
Formula:C₈H₄BrFN₂O₂
MDL Number:MFCD27981294
:259.032 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
Frequently Bought Together