2-Fluoro-5-(trifluoromethyl)pyrimidine 98%
Catalog # APOSPC902938-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:166.076 g/mol
Storage Temperature:Ambient
CAS Number:1214344-68-9
Formula:C₅H₂F₄N₂
MDL Number:MFCD13185662