
4-Methyl-3-(trifluoromethyl)phenylboronic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:947533-94-0
:
Formula:C₈H₈BF₃O₂
MDL Number:MFCD11042431
:203.954 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together