Order Entry
Czech Republic
ContactUsLinkComponent
2-Ethoxy-5-fluorobenzonitrile 96%
2-Ethoxy-5-fluorobenzonitrile 96%
Catalog # APOSPC901613-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Ethoxy-5-fluorobenzonitrile 96%
Catalog # APOSPC901613-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :165.164 g/mol
Storage Temperature:Ambient
CAS Number:1158201-03-6
Formula:C₉H₈FNO
MDL Number:MFCD15201675