Order Entry
Czech Republic
ContactUsLinkComponent
Methyl 2,6-difluoro-3-hydroxybenzoate 98%
Methyl 2,6-difluoro-3-hydroxybenzoate 98%
Catalog # APOSPC901603-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2,6-difluoro-3-hydroxybenzoate 98%
Catalog # APOSPC901603-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :188.128 g/mol
Storage Temperature:Ambient
CAS Number:1214332-41-8
Formula:C₈H₆F₂O₃
MDL Number:MFCD14698033