Order Entry
Czech Republic
ContactUsLinkComponent
1-(2-Fluoro-5-methoxyphenyl)ethanone 96%
1-(2-Fluoro-5-methoxyphenyl)ethanone 96%
Catalog # APOSPC901592-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Fluoro-5-methoxyphenyl)ethanone 96%
Catalog # APOSPC901592-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :168.165 g/mol
Storage Temperature:Ambient
CAS Number:80309-38-2
Formula:C₉H₉FO₂
MDL Number:MFCD01074363