2-(Cyclopropylmethoxy)-5-fluorobenzonitrile 95%
Catalog # APOSPC900219-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:191.202 g/mol
Storage Temperature:Ambient
CAS Number:1342025-16-4
Formula:C₁₁H₁₀FNO
MDL Number:MFCD18860667