2-Methyl-4-oxo-4-[3-(trifluoromethyl)phenyl]butanoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:66549-17-5
:
Formula:C₁₂H₁₁F₃O₃
MDL Number:MFCD04972251
:260.209 g/mol
Storage Temperature:Ambient
Frequently Bought Together