

4,5-Dichloroperfluoro(2,2-dimethyl-1,3-dioxolane)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:60644-92-0
:
Formula:C₅Cl₂F₈O₂
MDL Number:MFCD01937893
:314.946 g/mol
Boiling Pt:88…90 °C
Density:1.587 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1992
ADR:3+6.1,III
:GHS02
:GHS07
Frequently Bought Together