
4-Bromo-2-(trifluoromethyl)-1H-benzimidazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6587-23-1
:
Formula:C₈H₄BrF₃N₂
MDL Number:MFCD01658374
:265.03 g/mol
Melting Pt:154…156 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together