Order Entry
Czech Republic
ContactUsLinkComponent
3-(2-Aminoethyl)-5-fluoro-2-pyridin-3-yl-1H-indole hydrochloride
3-(2-Aminoethyl)-5-fluoro-2-pyridin-3-yl-1H-indole hydrochloride
Catalog # APOSPC8682-500MG
Supplier:  Apollo Scientific
CAS Number:  
3-(2-Aminoethyl)-5-fluoro-2-pyridin-3-yl-1H-indole hydrochloride
Catalog # APOSPC8682-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :291.751 g/mol
Storage Temperature:Ambient
CAS Number:1049787-77-0
Formula:C₁₅H₁₄FN₃·ClH
MDL Number:MFCD06245391