Order Entry
Czech Republic
ContactUsLinkComponent
2-[5-Fluoro-2-(pyridin-2-yl)-1H-indol-3-yl]ethylamine hydrochloride
2-[5-Fluoro-2-(pyridin-2-yl)-1H-indol-3-yl]ethylamine hydrochloride
Catalog # APOSPC8681-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-[5-Fluoro-2-(pyridin-2-yl)-1H-indol-3-yl]ethylamine hydrochloride
Catalog # APOSPC8681-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :291.751 g/mol
Storage Temperature:Ambient
CAS Number:1049787-50-9
Formula:C₁₅H₁₄FN₃·ClH
MDL Number:MFCD06245390