Pentafluorophenyl 3-(1,3-dioxolan-2-yl)thiophene-2-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:910037-02-4
:
Formula:C₁₄H₇F₅O₄S
MDL Number:MFCD09702392
:366.26 g/mol
Melting Pt:55.5…59.5 °C
Storage Temperature:Ambient
Frequently Bought Together