3-Amino-2-[4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl]pyridine hydrochloride 97%
Catalog # APOSPC8426-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:394.222 g/mol
Melting Pt:200…210 °C
Storage Temperature:Ambient
CAS Number:317822-49-4
Formula:C₁₅H₁₅ClF₃N₅·ClH
MDL Number:MFCD00793058