
4-Chloro-2,6-difluorophenylacetonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:537033-53-7
:
Formula:C₈H₄ClF₂N
MDL Number:MFCD03094465
:187.574 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,II
:GHS06
Frequently Bought Together