
3-(Trifluoromethoxy)benzene-1,2-diamine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:113638-49-6
:
Formula:C₇H₇F₃N₂O
MDL Number:MFCD16628051
:192.138 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together