Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tris(perfluorononyl)-s-triazine
Tris(perfluorononyl)-s-triazine 95%
Tris(perfluorononyl)-s-triazine 95%
Tris(perfluorononyl)-s-triazine 95%
APOSPC7912-1G
:
Apollo Scientific
:
Tris(perfluorononyl)-s-triazine 95%
APOSPC7912-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
1485.25 g/mol
:
82…84 °C
:
GHS07
:
Ambient
:
57101-59-4
:
C₃₀F₅₇N₃
:
MFCD00042440