
1,1-Dimethyl-2-fluoroethylamine hydrochloride
Catalog # APOSPC7908-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:127.588 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:112433-51-9
Formula:C₄H₁₀FN·ClH
MDL Number:MFCD07779327