
6-Methyl-2-(trifluoromethyl)-1H-benzimidazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:399-77-9
:
Formula:C₉H₇F₃N₂
MDL Number:MFCD00457964
:200.16 g/mol
Melting Pt:185…186 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together